Assay Details
| Gene Symbol: ankrd54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002578.1 | NP_001002578.1 | 7 - 8 | 1031 | 64 | |
| GenBank mRNA | BC076214.1 | - | 7 - 8 | 1031 | 64 | |
| BC092933.1 | - | 7 - 8 | 910 | 64 | ||
| BC096993.1 | - | 7 - 8 | 1106 | 64 | ||
More Information
Set Membership: |
|

ankrd54
Genomic Map