Assay Details
| Gene Symbol: psmg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002714.1 | NP_001002714.1 | 1 - 2 | 205 | 68 | |
| GenBank mRNA | BC076460.1 | - | 1 - 2 | 205 | 68 | |
More Information
Set Membership: |
|

psmg1
Genomic Map