Assay Details
| Gene Symbol: barx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024949.1 | NP_001020120.1 | 1 - 2 | 540 | 69 | |
| GenBank mRNA | AY596188.1 | - | 1 - 2 | 378 | 69 | |
| BC095353.1 | - | 1 - 2 | 540 | 69 | ||
| BC133991.1 | - | 1 - 2 | 544 | 69 | ||
| BC162220.1 | - | 1 - 2 | 251 | 69 | ||
| BC162228.1 | - | 1 - 2 | 251 | 69 | ||
| BC165106.1 | - | 1 - 2 | 243 | 69 | ||
More Information
Set Membership: |
|

barx1
Genomic Map