Assay Details
| Gene Symbol: barx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024949.1 | NP_001020120.1 | 2 - 3 | 820 | 89 | |
| GenBank mRNA | AY596188.1 | - | 2 - 3 | 658 | 89 | |
| BC095353.1 | - | 2 - 3 | 820 | 89 | ||
| BC133991.1 | - | 2 - 3 | 824 | 89 | ||
| BC162220.1 | - | 2 - 3 | 531 | 89 | ||
| BC162228.1 | - | 2 - 3 | 531 | 89 | ||
| BC165106.1 | - | 2 - 3 | 523 | 89 | ||
More Information
Set Membership: |
|

barx1
Genomic Map