Assay Details
| Gene Symbol: pknox1.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131891.3 | NP_571966.3 | 10 - 11 | 1255 | 91 | |
| XM_009304649.3 | XP_009302924.1 | 10 - 11 | 1483 | 91 | ||
| XM_017357727.2 | XP_017213216.1 | 9 - 10 | 1479 | 91 | ||
| GenBank mRNA | AF382393.1 | - | 9 - 10 | 1188 | 91 | |
| AY052752.1 | - | 9 - 10 | 1188 | 91 | ||
| BC155781.1 | - | 10 - 11 | 1184 | 91 | ||
More Information
Set Membership: |
|

pknox1.1
Genomic Map