Assay Details
| Gene Symbol: pknox1.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131891.3 | NP_571966.3 | 4 - 5 | 504 | 105 | |
| XM_009304649.3 | XP_009302924.1 | 4 - 5 | 732 | 105 | ||
| XM_017357727.2 | XP_017213216.1 | 3 - 4 | 728 | 105 | ||
| GenBank mRNA | AF382393.1 | - | 3 - 4 | 437 | 105 | |
| AY052752.1 | - | 3 - 4 | 437 | 105 | ||
| BC054667.1 | - | 4 - 5 | 436 | 105 | ||
| BC155781.1 | - | 4 - 5 | 433 | 105 | ||
| Gene Symbol: pknox1.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AL924758.1 | - | 2 - 3 | 294 | 105 | |
More Information
Set Membership: |
|

pknox1.1
Genomic Map