Assay Details
| Gene Symbol: pknox1.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131891.3 | NP_571966.3 | 7 - 8 | 896 | 87 | |
| XM_009304649.3 | XP_009302924.1 | 7 - 8 | 1124 | 87 | ||
| XM_017357727.2 | XP_017213216.1 | 6 - 7 | 1120 | 87 | ||
| GenBank mRNA | AF382393.1 | - | 6 - 7 | 829 | 87 | |
| AY052752.1 | - | 6 - 7 | 829 | 87 | ||
| BC054667.1 | - | 7 - 8 | 828 | 87 | ||
| BC155781.1 | - | 7 - 8 | 825 | 87 | ||
More Information
Set Membership: |
|

pknox1.1
Genomic Map