Assay Details
| Gene Symbol: pknox1.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131891.3 | NP_571966.3 | 9 - 10 | 1090 | 79 | |
| XM_009304649.3 | XP_009302924.1 | 9 - 10 | 1318 | 79 | ||
| XM_017357727.2 | XP_017213216.1 | 8 - 9 | 1314 | 79 | ||
| GenBank mRNA | AF382393.1 | - | 8 - 9 | 1023 | 79 | |
| AY052752.1 | - | 8 - 9 | 1023 | 79 | ||
| BC054667.1 | - | 9 - 10 | 1022 | 79 | ||
| BC155781.1 | - | 9 - 10 | 1019 | 79 | ||
More Information
Set Membership: |
|

pknox1.1
Genomic Map