Assay Details
| Gene Symbol: sesn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002660.1 | NP_001002660.1 | 4 - 5 | 670 | 89 | |
| XM_005160670.4 | XP_005160727.1 | 5 - 6 | 932 | 89 | ||
| XM_005160671.4 | XP_005160728.1 | 5 - 6 | 899 | 89 | ||
| XM_005160673.4 | XP_005160730.1 | 4 - 5 | 734 | 89 | ||
| GenBank mRNA | BC076550.1 | - | 4 - 5 | 670 | 89 | |
| DQ858215.1 | - | 4 - 5 | 579 | 89 | ||
More Information
Set Membership: |
|

sesn1
Genomic Map