Assay Details
| Gene Symbol: ccpg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001351662.1 | NP_001338591.1 | 228 | 55 | ||
| GenBank mRNA | BC085421.1 | - | 227 | 55 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ccpg1| Gene Symbol: ccpg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001351662.1 | NP_001338591.1 | 228 | 55 | ||
| GenBank mRNA | BC085421.1 | - | 227 | 55 | ||
Set Membership: |
|