Assay Details
| Gene Symbol: epyc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017903.4 | NP_001017903.3 | 7 - 8 | 1873 | 76 | |
| NM_001348483.1 | NP_001335412.1 | 5 - 6 | 905 | 76 | ||
| XM_005164743.4 | XP_005164800.1 | 8 - 9 | 2088 | 76 | ||
| XM_009300279.3 | XP_009298554.1 | 8 - 9 | 2085 | 76 | ||
| XM_021475014.1 | XP_021330689.1 | 5 - 6 | 908 | 76 | ||
| GenBank mRNA | BC092942.1 | - | 5 - 6 | 721 | 76 | |
| BC133975.1 | - | 5 - 6 | 730 | 76 | ||
| CU468782.1 | - | 4 - 5 | 692 | 76 | ||
| DQ224358.1 | - | 7 - 8 | 1869 | 76 | ||
More Information
Set Membership: |
|

epyc
Genomic Map