Assay Details
| Gene Symbol: cpb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020703.2 | NP_001018539.2 | 1 - 2 | 101 | 137 | |
| GenBank mRNA | BC095834.1 | - | 1 - 2 | 101 | 137 | |
| BC152600.1 | - | 1 - 2 | 102 | 137 | ||
More Information
Set Membership: |
|

cpb2
Genomic Map