Assay Details
Gene Symbol: selenot2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098487.2 | NP_001091957.2 | 2 - 3 | 420 | 99 | |
GenBank mRNA | AY216588.1 | - | 2 - 3 | 343 | 99 | |
BC065465.1 | - | 2 - 3 | 391 | 99 | ||
BC095658.1 | - | 2 - 3 | 99 | |||
CD285269.1 | - | 2 - 3 | 420 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
