Assay Details
| Gene Symbol: ap2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199919.2 | NP_956213.2 | 14 - 15 | 2108 | 54 | |
| GenBank mRNA | BC049138.1 | - | 14 - 15 | 2047 | 54 | |
| BC066566.1 | - | 14 - 15 | 2108 | 54 | ||
| BC165229.1 | - | 13 - 14 | 2024 | 54 | ||
More Information
Set Membership: |
|

ap2b1
Genomic Map