Assay Details
| Gene Symbol: pnpla3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002338.1 | NP_001002338.1 | 5 - 6 | 789 | 68 | |
| GenBank mRNA | BC075928.1 | - | 5 - 6 | 789 | 68 | |
More Information
Set Membership: |
|

pnpla3
Genomic Map