Assay Details
Gene Symbol: polr1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003888.1 | NP_001003888.1 | 1 - 2 | 45 | 65 | |
XM_005157141.4 | XP_005157198.1 | 1 - 2 | 86 | 65 | ||
GenBank mRNA | AY648775.1 | - | 1 - 2 | 45 | 65 | |
BC115138.1 | - | 1 - 2 | 45 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Molecular Function -



Gene Ontology Categories:
