Assay Details
Gene Symbol: polr1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003888.1 | NP_001003888.1 | 3 - 4 | 230 | 76 | |
XM_005157141.4 | XP_005157198.1 | 3 - 4 | 267 | 76 | ||
GenBank mRNA | AY648775.1 | - | 3 - 4 | 230 | 76 | |
BC115138.1 | - | 3 - 4 | 230 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Molecular Function -



Gene Ontology Categories:
