Assay Details
| Gene Symbol: hsdl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008607.1 | NP_001008607.1 | 2 - 3 | 855 | 79 | |
| GenBank mRNA | BC086821.1 | - | 2 - 3 | 855 | 79 | |
More Information
Set Membership: |
|

hsdl1
Genomic Map