Assay Details
| Gene Symbol: lin37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017888.1 | NP_001017888.1 | 7 - 8 | 541 | 76 | |
| GenBank mRNA | BC092956.1 | - | 7 - 8 | 541 | 76 | |
More Information
Set Membership: |
|

lin37
Genomic Map