Assay Details
| Gene Symbol: setdb1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077277.3 | NP_001070745.3 | 6 - 7 | 1153 | 80 | |
| XM_005158013.4 | XP_005158070.2 | 8 - 9 | 1364 | 80 | ||
| XM_005158015.4 | XP_005158072.2 | 8 - 9 | 1364 | 80 | ||
| XM_005158016.4 | XP_005158073.2 | 6 - 7 | 1163 | 80 | ||
| GenBank mRNA | BC124601.1 | - | 6 - 7 | 1096 | 80 | |
| BC165176.1 | - | 5 - 6 | 940 | 80 | ||
More Information
Set Membership: |
|

setdb1b
Genomic Map