Assay Details
| Gene Symbol: setdb1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077277.3 | NP_001070745.3 | 8 - 9 | 1407 | 69 | |
| XM_005158013.4 | XP_005158070.2 | 10 - 11 | 1618 | 69 | ||
| XM_005158015.4 | XP_005158072.2 | 10 - 11 | 1618 | 69 | ||
| XM_005158016.4 | XP_005158073.2 | 8 - 9 | 1417 | 69 | ||
| GenBank mRNA | BC124601.1 | - | 8 - 9 | 1350 | 69 | |
| BC165176.1 | - | 7 - 8 | 1194 | 69 | ||
| DQ358103.1 | - | 1 - 2 | 46 | 69 | ||
More Information
Set Membership: |
|

setdb1b
Genomic Map