Assay Details
| Gene Symbol: stpg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077347.1 | NP_001070815.1 | 721 | 71 | ||
| GenBank mRNA | BC124784.1 | - | 721 | 71 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

stpg2| Gene Symbol: stpg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077347.1 | NP_001070815.1 | 721 | 71 | ||
| GenBank mRNA | BC124784.1 | - | 721 | 71 | ||
Set Membership: |
|