Assay Details
| Gene Symbol: znf711 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105523.2 | NP_001098993.2 | 3 - 4 | 440 | 68 | |
| NM_001365610.1 | NP_001352539.1 | 3 - 4 | 440 | 68 | ||
| GenBank mRNA | EU122381.1 | - | 3 - 4 | 376 | 68 | |
More Information
Set Membership: |
|

znf711
Genomic Map