Assay Details
| Gene Symbol: sim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131836.1 | NP_571911.1 | 716 | 58 | ||
| GenBank mRNA | AF363019.1 | - | 716 | 58 | ||
| BC065360.1 | - | 695 | 58 | |||
More Information
Set Membership: |
|

sim2
Genomic Map