Assay Details
| Gene Symbol: zcchc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198370.1 | NP_938184.1 | 3 - 4 | 600 | 86 | |
| GenBank mRNA | BC049028.1 | - | 3 - 4 | 600 | 86 | |
More Information
Set Membership: |
|

zcchc9
Genomic Map