Assay Details
| Gene Symbol: akap17a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200682.2 | NP_956976.2 | 2 - 3 | 388 | 102 | |
| GenBank mRNA | BC058319.1 | - | 2 - 3 | 406 | 102 | |
| BC066673.1 | - | 2 - 3 | 388 | 102 | ||
More Information
Set Membership: |
|

akap17a
Genomic Map