Assay Details
| Gene Symbol: stoml2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201031.1 | NP_957325.1 | 7 - 8 | 867 | 66 | |
| GenBank mRNA | BC055126.1 | - | 7 - 8 | 867 | 66 | |
More Information
Set Membership: |
|

stoml2
Genomic Map