Assay Details
| Gene Symbol: crsp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213059.1 | NP_998224.1 | 2 - 3 | 391 | 100 | |
| GenBank mRNA | AY034615.1 | - | 2 - 3 | 370 | 100 | |
| BC045285.1 | - | 2 - 3 | 370 | 100 | ||
| BC045849.1 | - | 2 - 3 | 391 | 100 | ||
More Information
Set Membership: |
|

crsp7
Genomic Map