Assay Details
| Gene Symbol: znf330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213373.1 | NP_998538.1 | 4 - 5 | 319 | 86 | |
| GenBank mRNA | BC046062.1 | - | 4 - 5 | 319 | 86 | |
| BC071535.1 | - | 4 - 5 | 293 | 86 | ||
More Information
Set Membership: |
|

znf330
Genomic Map