Assay Details
| Gene Symbol: chst12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213547.2 | NP_998712.2 | 1 | 1211 | 73 | |
| GenBank mRNA | AB122017.1 | - | 1 | 1250 | 73 | |
| BC162067.1 | - | 1 | 1211 | 73 | ||
| BC162078.1 | - | 1 | 1211 | 73 | ||
More Information
Set Membership: |
|

chst12a
Genomic Map