Assay Details
| Gene Symbol: manea | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004566.2 | NP_001004566.2 | 3 - 4 | 646 | 106 | |
| XM_021467149.1 | XP_021322824.1 | 2 - 3 | 680 | 106 | ||
| GenBank mRNA | BC081681.1 | - | 2 - 3 | 639 | 106 | |
| EH485176.1 | - | 3 - 4 | 671 | 106 | ||
More Information
Set Membership: |
|

manea
Genomic Map