Assay Details
| Gene Symbol: timmdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017828.1 | NP_001017828.1 | 4 - 5 | 550 | 82 | |
| GenBank mRNA | BC092788.1 | - | 4 - 5 | 550 | 82 | |
More Information
Set Membership: |
|

timmdc1
Genomic Map