Assay Details
| Gene Symbol: hsd17b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040188.1 | NP_001035278.1 | 2 - 3 | 405 | 68 | |
| XM_005163177.4 | XP_005163234.1 | 2 - 3 | 1034 | 68 | ||
| GenBank mRNA | BC090448.1 | - | 2 - 3 | 343 | 68 | |
| DQ364628.1 | - | 2 - 3 | 405 | 68 | ||
More Information
Set Membership: |
|

hsd17b2
Genomic Map