Assay Details
| Gene Symbol: pcdh8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131209.2 | NP_571284.2 | 1 - 2 | 2449 | 131 | |
| GenBank mRNA | AF042191.1 | - | 1 - 2 | 2468 | 131 | |
More Information
Set Membership: |
|

pcdh8
Genomic Map