Assay Details
| Gene Symbol: klhl18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199830.1 | NP_956124.1 | 3 - 4 | 501 | 58 | |
| GenBank mRNA | BC045319.1 | - | 3 - 4 | 501 | 58 | |
More Information
Set Membership: |
|

klhl18
Genomic Map