Assay Details
| Gene Symbol: usp44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200257.1 | NP_956551.1 | 2 - 3 | 301 | 66 | |
| GenBank mRNA | BC048060.1 | - | 2 - 3 | 301 | 66 | |
| BC164039.1 | - | 1 - 2 | 217 | 66 | ||
More Information
Set Membership: |
|

usp44
Genomic Map