Assay Details
| Gene Symbol: map1lc3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200298.2 | NP_956592.1 | 1 - 2 | 76 | 61 | |
| GenBank mRNA | AB264295.1 | - | 1 - 2 | 59 | 61 | |
| AY398373.1 | - | 1 - 2 | 76 | 61 | ||
| BC049489.1 | - | 1 - 2 | 91 | 61 | ||
More Information
Set Membership: |
|

map1lc3c
Genomic Map