Assay Details
| Gene Symbol: psma4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214697.1 | NP_999862.1 | 8 - 9 | 716 | 67 | |
| GenBank mRNA | BC045970.1 | - | 8 - 9 | 716 | 67 | |
| BC164668.1 | - | 6 - 7 | 664 | 67 | ||
More Information
Set Membership: |
|

psma4
Genomic Map