Assay Details
| Gene Symbol: dctn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002220.1 | NP_001002220.1 | 1 - 2 | 128 | 102 | |
| GenBank mRNA | BC072726.1 | - | 1 - 2 | 128 | 102 | |
More Information
Set Membership: |
|

dctn3
Genomic Map