Assay Details
| Gene Symbol: negr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003851.1 | NP_001003851.1 | 4 - 5 | 844 | 73 | |
| XM_009302511.3 | XP_009300786.1 | 4 - 5 | 999 | 73 | ||
| GenBank mRNA | AF241637.1 | - | 4 - 5 | 844 | 73 | |
| BC080221.1 | - | 4 - 5 | 780 | 73 | ||
| CU458871.1 | - | 4 - 5 | 676 | 73 | ||
More Information
Set Membership: |
|

negr1
Genomic Map