Assay Details
Gene Symbol: magi1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007063.2 | NP_001007064.2 | 17 - 18 | 3536 | 112 | |
XM_005155829.4 | XP_005155886.1 | 17 - 18 | 3570 | 112 | ||
XM_005155830.4 | XP_005155887.1 | 17 - 18 | 3564 | 112 | ||
XM_005155834.4 | XP_005155891.1 | 6 - 7 | 1174 | 112 | ||
XM_005155835.4 | XP_005155892.1 | 6 - 7 | 1121 | 112 | ||
XM_009305951.3 | XP_009304226.1 | 17 - 18 | 3552 | 112 | ||
XM_009305953.3 | XP_009304228.1 | 16 - 17 | 3435 | 112 | ||
XM_009305956.3 | XP_009304231.1 | 17 - 18 | 3571 | 112 | ||
XM_021479602.1 | XP_021335277.1 | 16 - 17 | 3534 | 112 | ||
GenBank mRNA | AY465352.1 | - | 17 - 18 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
