Assay Details
| Gene Symbol: exosc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017585.1 | NP_001017585.1 | 5 - 6 | 517 | 73 | |
| GenBank mRNA | BC092997.1 | - | 5 - 6 | 517 | 73 | |
| BC152123.1 | - | 5 - 6 | 513 | 73 | ||
More Information
Set Membership: |
|

exosc7
Genomic Map