Assay Details
| Gene Symbol: dyrk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044833.2 | NP_001038298.1 | 2 - 3 | 508 | 76 | |
| XM_005164640.4 | XP_005164697.1 | 2 - 3 | 393 | 76 | ||
| XM_005164641.4 | XP_005164698.1 | 2 - 3 | 267 | 76 | ||
| GenBank mRNA | BC139631.1 | - | 2 - 3 | 508 | 76 | |
More Information
Set Membership: |
|

dyrk2
Genomic Map