Assay Details
| Gene Symbol: hs3st2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080608.1 | NP_001074077.1 | 1 - 2 | 529 | 86 | |
| GenBank mRNA | BC162784.1 | - | 1 - 2 | 558 | 86 | |
| DQ812986.1 | - | 1 - 2 | 529 | 86 | ||
More Information
Set Membership: |
|

hs3st2
Genomic Map