Assay Details
| Gene Symbol: si:ch211-214j8.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098190.1 | NP_001091660.1 | 15 - 16 | 1775 | 71 | |
| GenBank mRNA | BC171342.1 | - | 15 - 16 | 1775 | 71 | |
| BC171346.1 | - | 15 - 16 | 1775 | 71 | ||
More Information
Set Membership: |
|

si:ch211-214j8.1
Genomic Map