Assay Details
| Gene Symbol: rpl12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201584.4 | NP_963878.4 | 4 - 5 | 413 | 77 | |
| GenBank mRNA | AY648813.1 | - | 4 - 5 | 410 | 77 | |
| BC049061.1 | - | 4 - 5 | 413 | 77 | ||
| BC165413.1 | - | 4 - 5 | 329 | 77 | ||
More Information
Set Membership: |
|

rpl12
Genomic Map