Assay Details
| Gene Symbol: ahsa1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212602.1 | NP_997767.1 | 6 - 7 | 852 | 78 | |
| GenBank mRNA | BC055523.1 | - | 6 - 7 | 852 | 78 | |
| BC068225.1 | - | 6 - 7 | 746 | 78 | ||
| BC164374.1 | - | 6 - 7 | 725 | 78 | ||
More Information
Set Membership: |
|

ahsa1b
Genomic Map