Assay Details
| Gene Symbol: agmo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212883.2 | NP_998048.2 | 2 - 3 | 360 | 61 | |
| GenBank mRNA | BC066626.1 | - | 2 - 3 | 350 | 61 | |
| CK028737.1 | - | 2 - 3 | 255 | 61 | ||
More Information
Set Membership: |
|

agmo
Genomic Map