Assay Details
| Gene Symbol: fam32a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002203.1 | NP_001002203.1 | 2 - 3 | 218 | 74 | |
| GenBank mRNA | BC072708.1 | - | 2 - 3 | 218 | 74 | |
More Information
Set Membership: |
|

fam32a
Genomic Map