Assay Details
| Gene Symbol: nosip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007434.1 | NP_001007435.1 | 5 - 6 | 821 | 68 | |
| XM_005164022.4 | XP_005164079.1 | 6 - 7 | 661 | 68 | ||
| XM_009299536.3 | XP_009297811.1 | 6 - 7 | 719 | 68 | ||
| GenBank mRNA | BC085409.1 | - | 5 - 6 | 821 | 68 | |
| BC164074.1 | - | 5 - 6 | 580 | 68 | ||
More Information
Set Membership: |
|

nosip
Genomic Map